Learn More
Newcomer Supply Formalex Green Formalin Neutralizer, 1 gallon cube for drain disposal.
SDP
Catalog No. NC9683865
Encompass
Liquid formalin neutralizer that easily and safely neutralizes formalin waste to a non-hazardous, non-toxic solution for sink disposal. The improved formula eliminates any sludgy residue and does not require any pH adjustment. It contains no phosphates and is non-vapor forming. Use spray bottle for easy clean up of small countertop or bench spills!
Catalog No. NC9683865 Supplier Newcomer Supply Supplier No. ABFX01
Liquid formalin neutralizer that easily and safely neutralizes formalin waste to a non-hazardous, non-toxic solution for sink disposal. The improved formula eliminates any sludgy residue and does not require any pH adjustment. It contains no phosphates and is non-vapor forming. Use spray bottle for easy clean up of small countertop or bench spills!