Learn More
Newcomer Supply Formalex Green Formalin Neutralizer, 2.5 gallon cube for drain disposal.
SDP
Supplier:  Newcomer Supply ABFX250
Encompass
Liquid formalin neutralizer that easily and safely neutralizes formalin waste to a non-hazardous, non-toxic solution for sink disposal. The improved formula eliminates any sludgy residue and does not require any pH adjustment. It contains no phosphates and is non-vapor forming. Use spray bottle for easy clean up of small countertop or bench spills!
Catalog No. NC9301915