Learn More
Electron Microscopy Sciences Formalin 10% Buffered in Phosphate, 6.9 to 7.1 pH at 25°C pH 1 LT
SDP
Catalog No. 50980501
Encompass_Preferred
Buffered with phosphate in accordance with the A.f.I.P. manual of Histology. Filtered and ready-to-use. pH 6.9 to 7.1 at 25 degree C
pH: 6.9 to 7.1 pH at 25°C
Catalog No. 50-980-501 Supplier Electron Microscopy Sciences Supplier No. 1574001
Buffered with phosphate in accordance with the A.f.I.P. manual of Histology. Filtered and ready-to-use. pH 6.9 to 7.1 at 25 degree C
pH: 6.9 to 7.1 pH at 25°C