Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formalin, 10%, MilliporeSigma™

Catalog No. MR0337982 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MR0337982 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MR0337982 Supplier MilliporeSigma Supplier No. R0337982

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

Synonym NBF Fixative

Specifications

Physical Form Liquid
Color Colorless
Quantity 4 L
Assay Percent Range 10%
Chemical Name or Material Formalin, 10%, Fixative
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.