Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ Formalin, 10% Neutral Buffered
GSA_VASDP

Catalog No. STLNB0507
Change view
Click to view available options
Capacity (Metric):
20 mL
40 mL
60 mL
90 mL
120 mL
180 mL
240 mL
500 mL
1000 mL
No. per Case:
4
6
50
72
96
960
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) No. per Case
STLNB0507 20 mL 96
STLNB0507B 20 mL 960
STLNB0115 40 mL 96
STLNB0230 60 mL 96
STLNB0345 90 mL 96
STLNB0460 120 mL 96
STLNB0690 180 mL 50
STLNB08120 240 mL 72
STLNB16240 500 mL 6
STLNB32480 1000 mL 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. STLNB0507 Supplier StatLab™ Supplier No. NB0507
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

10% Neutral Buffered Formalin prefilled in StatClick containers that feature an audible clicking sound when the leak-proof seal has been properly engaged, assuring accurate closure and maintaining specimen integrity during transport.

  • StatClick containers are ready-to-use and feature a pH level of 7.0.
  • This prefill is packed in a StatLab HistoPak carrier for easy distribution.
  • HistoPak containers are 95 kPa certified for transport in compliance with DOT and IATA standards for leak-resistant transportation.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Pre-filled Formalin Container
Closure Included Yes
Graduated No
Material Polypropylene
Shape Cylindrical
Sterility Non-sterile
Autoclavable Not Autoclavable
Capacity (Metric) 20 mL
No. per Case 96
Fit for Purpose Certified Quality

Prefilled container

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.