Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formalin 10% Neutral Buffered, Vion Biosciences
SCI_ED

Catalog No. S02499 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 L
5 gal.
4 x 1 gal.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02499 4 x 1 gal.
S02501 500 mL
S02502 1 L
S02500 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
WARNING: This product can expose you to chemicals including Methanol and Formaldehyde, which are known to the State of California to cause cancer and birth defects or other reproductive harm.
Catalog No. S02499 Supplier Aldon Corporation Supplier No. VNFF0266CS/4
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Methanol and Formaldehyde, which are known to the State of California to cause cancer and birth defects or other reproductive harm.

This 10% neutral buffered formalin is a stable, neutral fixative used to ensure the long-term preservation of tissues as close to the living state as possible and provide clear, reproducible results.

  • This 10% neutral buffered formalin is a stable, neutral fixative used to ensure the long-term preservation of tissues as close to the living state as possible and provide clear, reproducible results.

Specifications

Boiling Point ~100°C (212°F in water)
Closure Type Screw Cap
Color Colorless
Content And Storage Store in a cool, well-ventilated area away from incompatible substances.
Packaging Type F Style UN Bottle
Concentration 10%
For Use With (Equipment) Microscope
Grade Histological
Product Line Vion Histology
Product Type Fixative
Quantity 4 x 1 gal.
Solution Type Formalin
Formulation 10% Neutral Buffered Formalin
For Use With (Application) Histology
pH 7
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.