Learn More
Newcomer Supply Formalin 10%, Phosphate Buffered, 1 gallon
SDP
Catalog No. NC9899672
Encompass
Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
Catalog No. NC9899672 Supplier Newcomer Supply Supplier No. 1090N
Shelf Life is 2 years from date of manufacture. Store at 15-30°C.