Learn More
Azer Scientific Formalin Absorbent Shipping and Grossing Pads
SDP
Catalog No. 22026237
Change view
Click to view available options
Type:
Formalin Absorbent Grossing Pad
Formalin Absorbent Shipping Pad
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-026-237 Formalin Absorbent Shipping Pad
22026238 Formalin Absorbent Grossing Pad
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-026-237 Supplier Azer Scientific Supplier No. ES86672
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Absorb and neutralize formalin and glutaraldehyde preventing hazardous vapours
  • Dense with excellent grossing surface
  • Designed to prevent dulling of blades
  • ES86671: Placement in a Biohazard bag (4 x 4 in. style)
  • ES86672: Grossing directly on the pad (8 x 11 in. style)
  • ES86673: Grossing directly on the pad (11 x 15 in. style)
Specifications
Type Formalin Absorbent Shipping Pad
For Use With (Application) Absorb and neutralize formalin and glutaraldehyde preventing hazardous vapors
Packaging Quantity 100/Cs.