Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formalin-Aceto-Alcohol (F-A-A Mixture), for Rapid Fixation of Tissue, Ricca Chemical
SDP

Catalog No. 321032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3210-32 1 L Natural Poly Bottle
3210-16 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3210-32 Supplier Ricca Chemical Company Supplier No. 321032
Only null left
Add to Cart
Add to Cart

Rapid Fixation of Tissue

Specifications

Physical Form Liquid
Chemical Name or Material Formalin-Aceto-Alcohol
Name Note F-A-A Mixture
CAS 7732-18-5, 64-17-5, 50-00-0, 67-56-1, 64-19-7
CAS Min % 43.0, 34.92, 17.29, 2.82
CAS Max % 45.0, 36.34, 18.0, 2.94
Packaging Natural Poly Bottle
Color Colorless
Quantity 1 L
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.