Learn More
Electron Microscopy Sciences Formalin-Acetone Fixative 500 ML
SDP
Catalog No. 5030061
Encompass_Preferred
Stains for hematologic elements. See reference Williams, W.j., Beutler, E., Erslev, A.J. and Rundles, R.W. Hematology, 2nd Ed. McGraw-Hill, YY, c 1977, pp 1627-1628.
Catalog No. 50-300-61 Supplier Electron Microscopy Sciences Supplier No. 2617301
Stains for hematologic elements. See reference Williams, W.j., Beutler, E., Erslev, A.J. and Rundles, R.W. Hematology, 2nd Ed. McGraw-Hill, YY, c 1977, pp 1627-1628.