Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Ascaris
SDPSCI_ED

Catalog No. S37687 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
10/Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S37687 10/Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37687 Supplier Carolina™ Supplier No. 224405
Add to Cart
Add to Cart

Includes

Tube of 10

Carolina™ Formalin Ascaris, is parasitic nematodealso known as roundworm.

  • Ascaris lumbricoides
  • Majority are females with very few, if any, males for comparison
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Carosafe is odorless fluid designed to minimize unpleasant
  • Tube of 10

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Quantity 10/Tube
Packaging Tube
Autoclavable Not Autoclavable
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.