Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Blue Crab
SDPSCI_ED

Catalog No. S72736 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
10/Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S72736 10/Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72736 Supplier Carolina™ Supplier No. 225222
Add to Cart
Add to Cart

Carolina™ Formalin Blue Crab is ideal for exploring the anatomy of an advanced crustacean in the classroom. Great for invertebrate and comparative anatomy classes.

  • Blue Crab (Callinectes sp.)
  • Great for studying crustacean anatomy
  • Preserved in formalin
  • Shipped in Carosafe™ Carolina's proprietary shipping and holding fluid
  • Carosafe minimizes student and educator exposure to formaldehyde

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Quantity 10/Pail
Packaging Pail
Autoclavable Not Autoclavable
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.