Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Bullfrogs
SDPSCI_ED

Catalog No. S37639 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Injection Type:
Double
Plain
Quantity:
1/Bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Injection Type Quantity
S37639 Double 1/Bag
S37638 Plain 1/Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S37639 Supplier Carolina™ Supplier No. 227121

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in introducing students to vertebrate anatomy. Carolina™ Formalin Bullfrogs are ideal for studying amphibian anatomy, adaptations, and body systems.

  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Carosafe is odorless fluid designed to minimize unpleasant odor of formaldehyde
  • INJECTIONS: Plain: no color injection; Double: red arteries, blue hepatic

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Double
Length (English) 5+ in.
Length (Metric) 12.7+ cm
Quantity 1/Bag
Packaging Bag
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.