Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Cats
SDPSCI_ED

Catalog No. p-7078171
Click to view available options
Injection Type:
Double
Plain
Single
Triple
Length (English):
14 to 18 in.
18+ in.
This item is not returnable. View return policy
This item is not returnable. View return policy

For use in advanced biology, anatomy and physiology classes. Carolina™ Formalin Cats are ideal dissection specimens.

  • Felis catus
  • Similar to human anatomy
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Carosafe is odorless fluid designed to minimize unpleasant odor of formaldehyde
  • INJECTIONS: Plain: n

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Double
Length (English) 18+ in.
Length (Metric) 45.7+ cm
Quantity 1/Bag
Packaging Bag
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.