Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Skinned Cats
SDPSCI_ED

Catalog No. S81620 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1/Bag
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S81620 1/Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S81620 Supplier Carolina™ Supplier No. 228102
This item is not returnable. View return policy

Formalin-preserved cats are ideal dissection specimens for advanced biology and anatomy and physiology classes.

  • Felis catus
  • Ideal specimen for studying advanced mammalian anatomy and physiology
  • Similar to human anatomy
  • Skin has been removed to save time and reveal underlying muscles
  • Preserved in formalin
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Carosafe™ is an odorless fluid designed to minimize the unpleasant odor of formaldehyde
  • INJECTIONS:
    • Plain: no color injection
    • Single: red arteries
    • Double: red arteries, blue systemic veins
    • Triple: red arteries, blue systemic veins, yellow hepatic system

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Double
Length (English) 18+ in.
Quantity 1/Bag
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.