Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Volu Sol Formalin Concentrate Buffered, 128 oz / 3.78 L, Volu-Sol™
SDP

Supplier:  Volu Sol VFC128

Encompass_Preferred

Makes five (5) gallons Formalin 10% Neutral Buffered

Catalog No. 50-420-850


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.