Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Cow Eyes
SDPSCI_ED

Catalog No. S37717 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Injection Type:
Plain
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Injection Type
S37717 Plain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37717 Supplier Carolina™ Supplier No. 228901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use in exploring anatomy of mammalian eye. Carolina™ Formalin Cow Eyes are similar to human eye

  • Large size allows easy dissection and examination
  • Preserved in formalin
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Includes One formalin preserved cow eye
Quantity 1 per bag
Packaging Bag
Autoclavable Not Autoclavable
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.