Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Crayfish
SDPSCI_ED

Catalog No. S37696 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Injection Type:
Plain
Single
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Injection Type
S37696 Single
S37695 Plain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S37696 Supplier Carolina™ Supplier No. 225292
Add to Cart
Add to Cart

For use in exploring relationship between structure, function, and environment using formalin-preserved crayfish. Carolina™ Formalin Crayfish is ideal invertebrate dissection specimen.

  • Pacifastacus sp.
  • Phylum Arthropoda, class Crustacea
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Carosafe is odorless fluid designed to minimize unpleasant odor of formaldehyde

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Single
Length (English) 4+ in.
Length (Metric) 10.1+ cm
Quantity 10/Bag
Packaging Bulk Bag
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.