Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Lamprey, Plain, 1 Per Bag
SDPSCI_ED

Catalog No. S72733 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1/Bag
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S72733 1/Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72733 Supplier Carolina™ Supplier No. 226601
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carolina™ Formalin Lamprey is for use in examining the anatomy of the jawless fish. Ideal for comparative anatomy classes and discussions about common ancestry and diversity.

  • Member of class Agnatha
  • Mature specimens
  • Characteristics: jawless vertebrate, paired gill openings, no paired fins, well developed eyes
  • Shipped in Carosafe™ Carolina's proprietary shipping and holding fluid
  • Carosafe minimizes student and educator exposure to formaldehyde

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Length (English) 15 to 20 in.
Quantity 1/Bag
Packaging Bag
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.