Learn More
Mopec Formalin Management Toolkit
SDP
Catalog No. 22038251 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Formalin Management Toolkit
Catalog No. Product Type
22-038-251 Formalin Management Toolkit
1 options
Catalog No. 22-038-251 Supplier Mopec Supplier No. BU005
Add to Cart
Add to Cart
Formalin and formalin neutralizing in one simple package. This toolkit includes the standard 2.5 gallon cube of formalin along with neutralizer pads for safer cutting and neutralizer powder for safer disposal of spent formalin.
The Toolkit includes:
  • (2) BE094-CS: Formaway Formalin. Neutralizing Pads, 11 x 15 in., 100 pack
  • (1) BG550-12: Formaway Formalin Neutralizing Powder, 12 pack
  • (4) BG520: Prefilled Formalin Cube, 2.5 gallons
We recommend ordering this toolkit every 2 months.
Specifications
Product Type Formalin Management Toolkit
Height (English) 3 in.
Length (English) 11 in.
Width (English) 15 in.