Learn More
Electron Microscopy Sciences NeutraGreen™ Formalin Neutralizer 5 GL
SDP
Catalog No. 501928593
Encompass_Preferred
Neutralizer that transforms waste formalin into an odorless, 100 percent nonhazardous material. Comes in liquid form with no powders to mix or dissolve. One of the fastest neutralizers available with a reaction time of only 2-3 hours. Third-party environmental data confirms NeutraGreen will neutralize waste formalin to a material that is non-toxic as well as non-hazardous.
Catalog No. 50-192-8593 Supplier Electron Microscopy Sciences Supplier No. 2414603
Neutralizer that transforms waste formalin into an odorless, 100 percent nonhazardous material. Comes in liquid form with no powders to mix or dissolve. One of the fastest neutralizers available with a reaction time of only 2-3 hours. Third-party environmental data confirms NeutraGreen will neutralize waste formalin to a material that is non-toxic as well as non-hazardous.