Learn More
Newcomer Supply FAN PAD-GL Large (Formalin Absorbing & Neutralizing Pad), 9" x 21", 30/case
SDP
Catalog No. NC0254690
Encompass
The FAN Pads (Formalin Absorbing Neutralizing Pads) are an excellent product for cleaning up spills, grossing on and using as a shelf liner where formalin specimens are stored or transported. The large FAN Pad is ideal for grossing formalin soaked tissue (e.g. uterus, breast and colon). It absorbs up to 500ml of 10% Formalin. Working Surface: 9" x 21".
Catalog No. NC0254690 Supplier Newcomer Supply Supplier No. ABFAN825
The FAN Pads (Formalin Absorbing Neutralizing Pads) are an excellent product for cleaning up spills, grossing on and using as a shelf liner where formalin specimens are stored or transported. The large FAN Pad is ideal for grossing formalin soaked tissue (e.g. uterus, breast and colon). It absorbs up to 500ml of 10% Formalin. Working Surface: 9" x 21".