Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Pig Heart
SDPSCI_ED

Catalog No. S37674 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1/Bag
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S37674 1/Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37674 Supplier Carolina™ Supplier No. 228561
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use in studying anatomy of cardiovascular system. Carolina™ Formalin Pig Heart is ideal for studying structure and function of heart.

  • Four-chambered mammalian heart
  • Like human heart, only larger
  • Easily trace blood flow through heart
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Quantity 1/Bag
Packaging Bag
Autoclavable Not Autoclavable
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.