Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Pig Kidney
SDPSCI_ED

Catalog No. S72739 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Injection Type:
Double
Triple
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Injection Type
S72739 Triple
S72738 Double
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S72739 Supplier Carolina™ Supplier No. 228591
Add to Cart
Add to Cart
This item is not returnable. View return policy

Carolina™ Formalin Pig Kidney is for use in enhancing study of urinary system. Similar to human kidney, only larger.

  • Mammalian kidney
  • For studying structure and function of kidney
  • Shipped in Carosafe™ Carolina's proprietary shipping and holding fluid
  • Carosafe is odorless fluid designed to minimize unpleasant odor of formaldehyde

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Triple
Quantity 1/Bag
Packaging Bag
Autoclavable Not Autoclavable
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.