Learn More
Electron Microscopy Sciences Formalin Prefilled HistoTainer, 20 ml
SDP
Catalog No. 50365871
Encompass_Preferred
The HistoTainer is half filled with 10% Neutral Buffered Formalin as a fixative. These ready-to-use prefilled containers are timer saving and make your work more efficient.
Catalog No. 50-365-871 Supplier Electron Microscopy Sciences Supplier No. 1574340
The HistoTainer is half filled with 10% Neutral Buffered Formalin as a fixative. These ready-to-use prefilled containers are timer saving and make your work more efficient.