Description
- Pump to draw fluid from tank
- Adjustable pressure controls
- System safety shutoff for gross leak (low pressure)
- Remote alarm system
- Electrical panel with indicator lights Includes containment platform with drain pan and leak sensor
Warranty and Services
1 Year Warranty
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Voltage | Alarm output (12 V and or NC/NO circuit) |
| Includes | Containment platform with drain pan and leak sensor |
| Product Type | Pump Station |