Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Rats
SDPSCI_ED

Catalog No. S37659 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Injection Type:
Double
Plain
Single
Type:
Preserved Specimen
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Injection Type Type
S37659 Plain Preserved Specimen
S37661 Double Preserved Specimen
S37660 Single Preserved Specimen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S37659 Supplier Carolina™ Supplier No. 228301
Add to Cart
Add to Cart

For use in lessons on mammalian anatomy. Carolina™ Formalin Rats have well-developed musculature and reproductive system.

  • Mature specimen
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Carosafe is odorless fluid designed to minimize unpleasant odor of formaldehyde
  • INJECTIONS: Plain: no color injection; Single: re

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Length (English) 7+ in.
Length (Metric) 17.7+ cm
Quantity 1/Bag
Packaging Bag
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.