Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Sea Cucumber
SDPSCI_ED

Catalog No. S37701 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
10/Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S37701 10/Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37701 Supplier Carolina™ Supplier No. 226110

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

For use in exploring anatomy of sea cucumber. Carolina™ Formalin Sea Cucumber is marine echinoderm.

  • Cucumaria sp.
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Carosafe is odorless fluid designed to minimize unpleasant odor of formaldehyde

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Length (English) 4 to 12 in., expanded
Length (Metric) 10.1 to 30.4 cm
Quantity 10/Pail
Packaging Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.