Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formalin Solution, Neutral Buffered, 10%, Sigma-Aldrich

Catalog No. 01258963 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-963 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-963 Supplier MilliporeSigma Supelco Supplier No. HT501850200L
Add to Cart
Add to Cart

10% NBF is well-suited for large throughput laboratories. It requires a relatively short period of fixation. It can be used for the long-term storage of tissue.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Microscope
Product Type Fixative
Quantity 200 L
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.