Learn More
Newcomer Supply Formalin Vial 10%, Phosphate Buffered, 20 ml vial, 10 ml fill, 100/case
SDP
Catalog No. NC2101888
Encompass
Shelf Life is 2 years from date of manufacture. Store at 15-30°C. Vials have an O-ring lid for secure seal.
This item is not returnable. View return policy
Catalog No. NC2101888 Supplier Newcomer Supply Supplier No. 1090A
Shelf Life is 2 years from date of manufacture. Store at 15-30°C. Vials have an O-ring lid for secure seal.