Learn More
Leica Biosystems D-Formalizer Powder - Aldehyde Neutralizer for Spent Formalin, 10 x 680 g
Catalog No. NC0283618
Encompass
D-Formalizer is a powdered aldehyde neutralizer for histology laboratories that treats spent formalin and 4% glutaraldehyde, producing a clear, non-odorous solution suitable for disposal. It simplifies chemical waste handling by neutralizing aldehydes and minimizing solids that can clog drains; one packet is formulated to treat approximately one gallon of used fixative.
  • Neutralizes spent formalin and 4% glutaraldehyde
  • Produces a clear solution free of solids and aldehyde odor
  • Simplifies hazardous waste handling and disposal
  • Single-packet dosing treats about one gallon of used fixative
  • Powdered form mixes easily for quick treatment
Catalog No. NC0283618 Supplier Leica Biosystems Supplier No. 3800753
D-Formalizer is a powdered aldehyde neutralizer for histology laboratories that treats spent formalin and 4% glutaraldehyde, producing a clear, non-odorous solution suitable for disposal. It simplifies chemical waste handling by neutralizing aldehydes and minimizing solids that can clog drains; one packet is formulated to treat approximately one gallon of used fixative.
  • Neutralizes spent formalin and 4% glutaraldehyde
  • Produces a clear solution free of solids and aldehyde odor
  • Simplifies hazardous waste handling and disposal
  • Single-packet dosing treats about one gallon of used fixative
  • Powdered form mixes easily for quick treatment