Learn More
Apexbio Technology LLC FORMAMIDE-100MG
SDP
Catalog No. 5000569497
Encompass_Preferred
Formamide (CAS No 75-12-7) is a versatile compound extensively used in biomedical research due to its ability to act as a solvent and its role in stabilizing nucleic acids Originating from carbonyl and amine it facilitates reactions involving hydrogen bond formation Its mechanism of action often involves interaction with nucleic acid structures influencing various biological pathways In vitro activity generally falls within the nanomolar to low micromolar range making it valuable for precise molecular studies Formamide is frequently employed in cell models and animal studies to explore genetic manipulation and gene expression effects It serves as a critical component in drug screening processes aiding in the evaluation of therapeutic candidates Experimental concentrations typically depend on the specific research design ensuring adaptability across diverse experiments
Catalog No. 50-005-69497 Supplier Apexbio Technology LLC Supplier No. C6876100
 Restricted Item
Add to Cart
Add to Cart
Formamide (CAS No 75-12-7) is a versatile compound extensively used in biomedical research due to its ability to act as a solvent and its role in stabilizing nucleic acids Originating from carbonyl and amine it facilitates reactions involving hydrogen bond formation Its mechanism of action often involves interaction with nucleic acid structures influencing various biological pathways In vitro activity generally falls within the nanomolar to low micromolar range making it valuable for precise molecular studies Formamide is frequently employed in cell models and animal studies to explore genetic manipulation and gene expression effects It serves as a critical component in drug screening processes aiding in the evaluation of therapeutic candidates Experimental concentrations typically depend on the specific research design ensuring adaptability across diverse experiments