Learn More
Medchemexpress LLC Formamide-13C | 51284-91-4 | MFCD00083968 | 99.9% | 46.03 | 13CH3NO | 100mg
SDP
Catalog No. 5000267852
Encompass_Preferred
Formamide-13C is the 13C-labeled Formamide (HY-Y0842) Formamide is an effective DNA denaturant that significantly reduces the stability of DNA in buffer solutions Formamide can also be used as a decalcifying agent for rat cardiac cells Additionally formamide is widely used as a solvent or chemical raw material for ion compounds resins and plasticizers [1][2][3][4][5][6][7]
Catalog No. 50-002-67852 Supplier Medchemexpress LLC Supplier No. HYY0842S100MG
Formamide-13C is the 13C-labeled Formamide (HY-Y0842) Formamide is an effective DNA denaturant that significantly reduces the stability of DNA in buffer solutions Formamide can also be used as a decalcifying agent for rat cardiac cells Additionally formamide is widely used as a solvent or chemical raw material for ion compounds resins and plasticizers [1][2][3][4][5][6][7]