Learn More
Medchemexpress LLC Formamide-15N | 1449-77-0 | MFCD00084213 | 99.5% | 46.03 g/mol | CH315NO | 100mg
SDP
Catalog No. 5000268837
Encompass_Preferred
Formamide-15N is the 15N labeled Formamide (HY-Y0842) Formamide is an effective DNA denaturant that significantly reduces the stability of DNA in buffer solutions Formamide can also be used as a decalcifying agent for rat cardiac cells Additionally formamide is widely used as a solvent or chemical raw material for ion compounds resins and plasticizers [1][2][3][4][5][6][7]
Catalog No. 50-002-68837 Supplier Medchemexpress LLC Supplier No. HYY0842S3100MG
Formamide-15N is the 15N labeled Formamide (HY-Y0842) Formamide is an effective DNA denaturant that significantly reduces the stability of DNA in buffer solutions Formamide can also be used as a decalcifying agent for rat cardiac cells Additionally formamide is widely used as a solvent or chemical raw material for ion compounds resins and plasticizers [1][2][3][4][5][6][7]