Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamide, Spectrophotometric Grade, ≥99%, Solstice

Catalog No. 60048450 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-450 2 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-450 Supplier Solstice Supplier No. 2958761L

missing translation for 'validMainframeMsg'

Spectrophotometric Grade,≥99%

  • Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 1°C to 3°C
CAS 75-12-7
Density 1.134g/mL at 25°C
Quantity 2 L
Flash Point 175°C (347°F)
Ignition Point 932°F
Packaging Glass Bottle
Refractive Index 1.447
Molecular Formula CH3NO
Vapor Pressure 0.08mmHg (20°C), 30mmHg (129°C)
Linear Formula HCONH2
MDL Number MFCD00007941
UN Number NONH for all modes of transport
Beilstein 505995
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
Boiling Point 210°C
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
SMILES NC=O
IUPAC Name formamide
Molecular Weight (g/mol) 45.04
PubChem CID 713
ChEBI CHEBI:48431
Formula Weight 45.04g/mol
Percent Purity ≥99%
Grade Spectrophotometry
Chemical Name or Material Formamide
Show More Show Less
Hazard Statement P201-P281-P308 + P313
Precautionary Statement H351-H360D-H373
EINECSNumber 200-842-0
RTECSNumber LQ0525000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.