Learn More
Bioworld Formamide, Deionized, 100 mL
Supplier:  Bioworld 406700081
Encompass_Preferred
It is a clear liquid amide derived from formic acid and can be used as cryoprotectant and gel-stabilizer. It is known to destabilize nucleic acid duplexes, making it useful for preparing hybridization buffers for RNA and DNA applications. Technical Information:Conductivity (µmhos): <100Abs. @ 280nm: <0.5Freezing Range: 1-3°Iron: <0.0005%Copper: <0.00001%Lead: <0.00005%ZInc: <0.00005%RNase/DNase: None DetectedAppearance: LiquidAtomic Number: Not available.Boiling Point : 210 °C(lit.)ChEBI : Not available.Crystal_Structure : Not available.Density : 1.134 g/mL at 25 °C(lit.)Electron Configuration : Not available.InChI : Not available.InChIKey : Not available.Ionization Energy : Not available.Melting Point: 2-3 °C(lit.)Molar Mass: Not available.NFPA 704: Not available.RTECS Number: LQ0525000Synonyms: Amide C1, Formic amide
Catalog No. 50-256-001