Learn More
Medchemexpress LLC Formamide (deionized) | 75-12-7 | MFCD00007941 | 100.0% | 45.04 | CH3NO | 500mL
SDP
Catalog No. 5000268629
Encompass_Preferred
Formamide deionized (Methanamide deionizde) is an effective DNA denaturant that significantly reduces the stability of DNA in buffer solutions Formamide deionized can also be used as a decalcifying agent for rat cardiac cells Additionally Formamide deionized is widely used as a solvent or chemical raw material for ion compounds resins and plasticizers[1][2][3][4][5][6][7]
Catalog No. 50-002-68629 Supplier Medchemexpress LLC Supplier No. HYY0842B500ML
Formamide deionized (Methanamide deionizde) is an effective DNA denaturant that significantly reduces the stability of DNA in buffer solutions Formamide deionized can also be used as a decalcifying agent for rat cardiac cells Additionally Formamide deionized is widely used as a solvent or chemical raw material for ion compounds resins and plasticizers[1][2][3][4][5][6][7]