Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Formamide, Deionized, OmniPur™, Calbiochem™,

Catalog No. 4610100ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-101-00ML 100 mL
46-505-00ML 500 mL
46-704-L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 46-101-00ML Supplier MilliporeSigma™ Supplier No. 4610100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Melting Point 2.9°C
Color Colorless
Density 1.13g/cm3 at 20°C
Quantity 100 mL
Flash Point 154.9°C (310.8°F), Open Cup
Odor Odorless
Physical Form Liquid
Boiling Point 209.9°C
Formula Weight 45
Percent Purity ≥99%
Grade Molecular Biology
Chemical Name or Material Formamide
Show More Show Less
Hazard Category H319: Causes serious eye irritation.
Hazard Statement H360D May damage the unborn child.
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.