Learn More
Alkali Scientific Formamide, Molecular Biology, 1 L
SDP
Catalog No. 5000661309
Encompass_Preferred
Formamide, Molecular Biology Grade, 1L, is a high-purity reagent essential in molecular biology applications. It is commonly used in nucleic acid research, particularly for DNA and RNA denaturation, sequencing, and hybridization. Formamide disrupts hydrogen bonding, aiding in the unwinding of double-stranded DNA and the denaturation of RNA. It is a key reagent for preparing samples for gel electrophoresis and for reverse transcription. The 1-liter bottle offers a larger supply for laboratories requiring substantial amounts of formamide for extended experimental runs. The molecular biology grade ensures high purity for reliable and reproducible results.
Catalog No. 50-006-61309 Supplier Alkali Scientific Supplier No. ORM003CP
Formamide, Molecular Biology Grade, 1L, is a high-purity reagent essential in molecular biology applications. It is commonly used in nucleic acid research, particularly for DNA and RNA denaturation, sequencing, and hybridization. Formamide disrupts hydrogen bonding, aiding in the unwinding of double-stranded DNA and the denaturation of RNA. It is a key reagent for preparing samples for gel electrophoresis and for reverse transcription. The 1-liter bottle offers a larger supply for laboratories requiring substantial amounts of formamide for extended experimental runs. The molecular biology grade ensures high purity for reliable and reproducible results.