Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamide, puriss. p.a., ACS Reagent, ≥99.5% (GC/T), Solstice

Catalog No. 60046961 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-961 1 L Glass Bottle
60-046-963 250 mL Glass Bottle
60-046-962 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-046-961 Supplier Solstice Supplier No. 476701L
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent,≥99.5% (GC/T)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

pH 4 to 5 (at 20°C)
Melting Point 1°C to 3°C
Color Colorless
Density 1.132g/mL at 20°C, 1.134g/mL at 25°C
Quantity 1 L
Flash Point 175°C (347°F)
Ignition Point 932°F
Odor Weak
Packaging Glass Bottle
Physical Form Liquid
Linear Formula HCONH2
Vapor Pressure 30mmHg (129°C), 0.08mmHg (20°C)
UN Number NONH for all modes of transport
Beilstein 505995
Boiling Point 210°C
Formula Weight 45.04g/mol
Percent Purity ≥99.5% (GC/T)
Chemical Name or Material Formamide
Show More Show Less
Hazard Statement P201-P260-P280-P308 + P313
RTECSNumber 200-842-0
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.