Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamide Solution NMR Reference Standard, 90% in DMSO-d6 (99.9 atom % D), NMR Tube Size: 5 mm 8 in., MilliporeSigma™ Supelco™

Catalog No. 111017112 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
0.7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7112 0.7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7112 Supplier MilliporeSigma Supelco Supplier No. 5513848IN
Add to Cart
Add to Cart

Specifications

Packaging NMR Tube Size: 5 mm 8 in.
Concentration 90% in DMSO- d 6 (99.9 atom% D)
Linear Formula CH3NO
MDL Number MFCD00007941
Quantity 0.7 mL
Formula Weight 45.04
Percent Purity 99% (CP)
Grade NMR Reference Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.