Learn More
Alkali Scientific Formamide, Super Pure, 100 Milliliters
SDP
Catalog No. 5000661304
Encompass_Preferred
Formamide, Super Pure, 100 Milliliters, is a high-purity reagent commonly used in molecular biology. Formamide is used in various laboratory applications, including DNA sequencing, hybridization, and protein synthesis. Its ability to disrupt hydrogen bonding makes it ideal for denaturing nucleic acids, enabling the separation of double-stranded DNA. It is also utilized in RNA hybridization to facilitate the denaturation of RNA-DNA hybrids. The super pure grade ensures minimal contamination, making it suitable for highly sensitive and accurate experimental procedures. This reagent is essential for nucleic acid research and other molecular biology applications requiring controlled conditions.
Catalog No. 50-006-61304 Supplier Alkali Scientific Supplier No. ORM001CP
Formamide, Super Pure, 100 Milliliters, is a high-purity reagent commonly used in molecular biology. Formamide is used in various laboratory applications, including DNA sequencing, hybridization, and protein synthesis. Its ability to disrupt hydrogen bonding makes it ideal for denaturing nucleic acids, enabling the separation of double-stranded DNA. It is also utilized in RNA hybridization to facilitate the denaturation of RNA-DNA hybrids. The super pure grade ensures minimal contamination, making it suitable for highly sensitive and accurate experimental procedures. This reagent is essential for nucleic acid research and other molecular biology applications requiring controlled conditions.