Learn More
Alkali Scientific Formamide, Super Pure, 500 Milliliters
SDP
Catalog No. 5000661547
Encompass_Preferred
Formamide, Super Pure, 500 Milliliters, is a high-purity chemical reagent widely used in molecular biology. Formamide is integral for processes like RNA and DNA hybridization, denaturation, and sequencing. In RNA analysis, formamide facilitates the separation of double-stranded RNA by disrupting hydrogen bonds. It is also utilized in reverse-transcription reactions and protein synthesis protocols. The super pure grade provides minimal impurities, ensuring accuracy in sensitive experiments. This 500mL bottle is an essential component in many laboratory applications, offering a convenient, larger volume for long-term projects in nucleic acid research.
Catalog No. 50-006-61547 Supplier Alkali Scientific Supplier No. ORM002CP
Formamide, Super Pure, 500 Milliliters, is a high-purity chemical reagent widely used in molecular biology. Formamide is integral for processes like RNA and DNA hybridization, denaturation, and sequencing. In RNA analysis, formamide facilitates the separation of double-stranded RNA by disrupting hydrogen bonds. It is also utilized in reverse-transcription reactions and protein synthesis protocols. The super pure grade provides minimal impurities, ensuring accuracy in sensitive experiments. This 500mL bottle is an essential component in many laboratory applications, offering a convenient, larger volume for long-term projects in nucleic acid research.