Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Formamide, Ultra High Purity 500 ML
SDP

Catalog No. 50980511
Encompass_Preferred

CAS #75-12-7 HCONH₂ F.W. 45.04
Assay: 99.9%

Glass Distilled. Exceeds A.C.S. specifications.

Catalog No. 50-980-511 Supplier Electron Microscopy Sciences Supplier No. 15747
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS #75-12-7 HCONH₂ F.W. 45.04
Assay: 99.9%

Glass Distilled. Exceeds A.C.S. specifications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.