Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formanilide, 99+%

Catalog No. AC158425000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158425000 500 g Plastic bottle
AC158420050 5 g Glass bottle
AC158421000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC158425000 Supplier Thermo Scientific Chemicals Supplier No. 158425000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 103-70-8 | C7H7NO | 121.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-70-8
MDL Number MFCD00003276
InChI Key DYDNPESBYVVLBO-UHFFFAOYSA-N
Synonym formanilide, formamide, n-phenyl, n-formylaniline, phenyl formamide, formamidobenzene, formylaniline, carbanilaldehyde, aniline, n-formyl, phenylformamide, n-phenyl-formamide
PubChem CID 7671
ChEBI CHEBI:42416
IUPAC Name N-phenylformamide
SMILES C1=CC=C(C=C1)NC=O

Specifications

Melting Point 46°C to 49°C
Color Beige-Gray to White
Density 1.14g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Molecular Formula C7H7NO
Linear Formula HCONHC6H5
Quantity 500 g
Beilstein 12, 230
Merck Index 15, 4266
Molecular Weight (g/mol) 121.139
Formula Weight 121.14
Percent Purity ≥99.0%
Physical Form Crystals
Chemical Name or Material Formanilide, 99+%
Show More Show Less
EINECSNumber 203-136-0
RTECSNumber BY1780000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.