Learn More
Beckman Coulter DNA Isolation From FFPE Tissue
Catalog No. NC9588771
Encompass
The Genomic DNA Isolation Kit from Beckman Coulter is designed to isolate genomic DNA from formalin-fixed, paraffin-embedded (FFPE) tissue sections. It uses a column-based method to purify DNA, removing cross-links, inhibitors, and contaminants. The kit includes a microtube pestle for lysing samples, Proteinase K for digestion, Lysis Buffer, Wash Buffers, Elution Buffer, and Spin Columns. It can process up to 4 x 25um sections per prep, yields up to 12 μg DNA per sample, and produces DNA ready for PCR, Southern blotting, and other applications. The streamlined procedure takes around 90 minutes.
Catalog No. NC9588771 Supplier Beckman Coulter Supplier No. A33341
The Genomic DNA Isolation Kit from Beckman Coulter is designed to isolate genomic DNA from formalin-fixed, paraffin-embedded (FFPE) tissue sections. It uses a column-based method to purify DNA, removing cross-links, inhibitors, and contaminants. The kit includes a microtube pestle for lysing samples, Proteinase K for digestion, Lysis Buffer, Wash Buffers, Elution Buffer, and Spin Columns. It can process up to 4 x 25um sections per prep, yields up to 12 μg DNA per sample, and produces DNA ready for PCR, Southern blotting, and other applications. The streamlined procedure takes around 90 minutes.