Learn More
Beckman Coulter FormaPure XL RNA Reagent Kit, 50 Preps
Catalog No. NC1745036
Encompass
RNA extraction from FFPE samples have been demonstrated using this kit. This method can be performed in 1.5 mL tube format or in a 96-well 1.2 mL plate format. There are enough materials in this kit for 50 preps.
Catalog No. NC1745036 Supplier Beckman Coulter Supplier No. C36000
RNA extraction from FFPE samples have been demonstrated using this kit. This method can be performed in 1.5 mL tube format or in a 96-well 1.2 mL plate format. There are enough materials in this kit for 50 preps.