Learn More
Roche Diagnostics Formate Bio HT, 200 tests
Supplier:  Roche Diagnostics 07705603001
Encompass
This product is intended for quantitative determination of formate (formic acid) in aqueous solutions using the Cedex Bio HT Analyzer.
Catalog No. NC1351992