Learn More
Medchemexpress LLC Formate dehydrogenase | 9028-85-7 | 100 U
SDP
Catalog No. 5000397429
Encompass_Preferred
Formate dehydrogenase is a class of oxidoreductases found widely in bacteria, fungi, plants, and animals. It catalyzes the reversible conversion between formic acid and carbon dioxide, involving redox reactions of the coenzyme NAD+/NADH or other electron carriers.
  • Sourced from Pseudomonas syringae
  • Improves resistance of Saccharomyces cerevisiae to formic acid and acetic acid
  • Enhances aluminum tolerance in tobacco
Catalog No. 50-003-97429 Supplier Medchemexpress LLC Supplier No. HYP2907100U
Formate dehydrogenase is a class of oxidoreductases found widely in bacteria, fungi, plants, and animals. It catalyzes the reversible conversion between formic acid and carbon dioxide, involving redox reactions of the coenzyme NAD+/NADH or other electron carriers.
  • Sourced from Pseudomonas syringae
  • Improves resistance of Saccharomyces cerevisiae to formic acid and acetic acid
  • Enhances aluminum tolerance in tobacco