Learn More
Sigma Aldrich Fine Chemicals Biosciences formate Dehydrogenase from
Catalog No. 502905498
Encompass
formateNAD oxidoreductaseformate dehydrogenase is part of the D-specific 2-hydroxy acid superfamily of dehydrogenases. It is a dimer of two identical subunits. The enzyme has a cysteine residue which is essential for its activity or structural integrity.
Catalog No. 50-290-5498 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10244678001
formateNAD oxidoreductaseformate dehydrogenase is part of the D-specific 2-hydroxy acid superfamily of dehydrogenases. It is a dimer of two identical subunits. The enzyme has a cysteine residue which is essential for its activity or structural integrity.